accessibility menu, dialog, popup

UserName

H3CH3CCH3OOHN(S)OONOOHNOOCH3H3CCH3

CAS RN 30189-36-7

IUPAC Name: 2,5-dioxopyrrolidin-1-yl (2S)-2,6-bis({[(tert-butoxy)carbonyl]amino})hexanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl (2S)-2,6-bis[(tert-butoxycarbonyl)amino]hexanoate
Molecular Weight (g/mol): 443.5
Molecular Formula: C20H33N3O8
InChi Key: IQVLXQGNLCPZCL-UGPWUYPHNA-N
SMILES: CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)