accessibility menu, dialog, popup

UserName

CAS RN 3003-84-7

IUPAC Name: 2-(chloromethyl)oxolane
Synonyms: 2-chloromethyl tetrahydrofuran
Molecular Weight (g/mol): 120.58
Molecular Formula: C5H9ClO
InChi Key: IVJLGIMHHWKRAN-UHFFFAOYNA-N
SMILES: ClCC1CCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results