accessibility menu, dialog, popup

UserName

CAS RN 3002-77-5

CH3NN

CAS RN 3002-77-5

IUPAC Name: 2-methyl-1,10-phenanthroline
Synonyms: 2-methyl-1,10-phenanthroline
Molecular Weight (g/mol): 194.24
Molecular Formula: C13H10N2
InChi Key: LQZDYPHFVGRHKD-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC3=CC=CN=C3C2=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results