accessibility menu, dialog, popup

UserName

CAS RN 29703-01-3

CsOOHO

CAS RN 29703-01-3

IUPAC Name: caesium(1+) hydrogen carbonate
Synonyms: caesium bicarbonate
Molecular Weight (g/mol): 193.92
Molecular Formula: CHCsO3
InChi Key: ZMCUDHNSHCRDBT-UHFFFAOYSA-M
SMILES: [Cs+].OC([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results