accessibility menu, dialog, popup

UserName

H3CCH3N

CAS RN 2961-47-9

IUPAC Name: 4-(nonan-5-yl)pyridine
Synonyms: 4-(nonan-5-yl)pyridine
Molecular Weight (g/mol): 205.35
Molecular Formula: C14H23N
InChi Key: ATCGHKBXRIOBDX-UHFFFAOYSA-N
SMILES: CCCCC(CCCC)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results