accessibility menu, dialog, popup

UserName

FFFFFFHO

CAS RN 29463-77-2

IUPAC Name: 1,1,1,3,3,3-hexafluoropropan-2-ol
Synonyms: hexafluoroisopropanol hexafluoroisopropyl alcohol 2H-hexafluoroisopropanol HFIP hexafluoro-2-propanol
Molecular Weight (g/mol): 168.04
Molecular Formula: C3H2F6O
InChi Key: BYEAHWXPCBROCE-UHFFFAOYSA-N
SMILES: OC(C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results