accessibility menu, dialog, popup

UserName

CAS RN 29342-44-7

H3CCH3CH3CH3Cl

CAS RN 29342-44-7

IUPAC Name: 2-chloro-2,5-dimethylhexane
Synonyms: 2-chloro-2,5-dimethylhexane
Molecular Weight (g/mol): 148.67
Molecular Formula: C8H17Cl
InChi Key: SVRDRJZPEYTXKX-UHFFFAOYSA-N
SMILES: CC(C)CCC(C)(C)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results