accessibility menu, dialog, popup

UserName

BaH3COH3CO

CAS RN 2914-19-4

IUPAC Name: barium(2+) diethanolate
Synonyms: barium(2+) bis(ethoxide)
Molecular Weight (g/mol): 227.45
Molecular Formula: C4H10BaO2
InChi Key: GYIWFHXWLCXGQO-UHFFFAOYSA-N
SMILES: [Ba++].CC[O-].CC[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results