CAS RN 29051-44-3
CAS RN 29051-44-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-Phenylnicotinic acid, 95%, Thermo Scientific™
CAS: 29051-44-3 Molecular Formula: C12H9NO2 Molecular Weight (g/mol): 199.209 MDL Number: MFCD03086128 InChI Key: DLFLQXUYRFIFOK-UHFFFAOYSA-N Synonym: 6-phenylnicotinic acid,3-pyridinecarboxylic acid, 6-phenyl,6-phenyl-nicotinic acid,2-phenyl pyridine-5-carboxylic acid,6-phenyl nicotinic acid,acmc-1cmxf,2-phenyl-5-carboxypyridine,3-carboxy-6-phenylpyridine,2-phenyl-5-pyridinecarboxylic acid,6-phenyl-3-pyridinecarboxylic acid PubChem CID: 120118 IUPAC Name: 6-phenylpyridine-3-carboxylic acid SMILES: C1=CC=C(C=C1)C2=NC=C(C=C2)C(=O)O