CAS RN 2902-65-0
IUPAC Name:
2-iodobenzene-1,3-dicarboxylic acid
Synonyms:
2-iodobenzene-1,3-dicarboxylic acid
Molecular Weight (g/mol):
292.03
Molecular Formula:
C8H5IO4
InChi Key:
PBIDRTDEJPAXHY-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=CC(C(O)=O)=C1I
1
–
1
of
1
results
1
2-Iodoisophthalic Acid 98.0+%, TCI America™
CAS: 2902-65-0 Molecular Formula: C8H5IO4 Molecular Weight (g/mol): 292.028 MDL Number: MFCD00156976 InChI Key: PBIDRTDEJPAXHY-UHFFFAOYSA-N PubChem CID: 320100 IUPAC Name: 2-iodobenzene-1,3-dicarboxylic acid SMILES: C1=CC(=C(C(=C1)C(=O)O)I)C(=O)O