accessibility menu, dialog, popup

UserName

H3CNNCH3CH3I

CAS RN 288627-94-1

IUPAC Name: 1-hexyl-2,3-dimethyl-1H-imidazol-3-ium iodide
Synonyms: 3-hexyl-1,2-dimethylimidazol-1-ium iodide
Molecular Weight (g/mol): 308.21
Molecular Formula: C11H21IN2
InChi Key: YYXZQUOJBJOARI-UHFFFAOYSA-M
SMILES: [I-].CCCCCCN1C=C[N+](C)=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results