CAS RN 28862-79-5
CAS RN 28862-79-5
IUPAC Name:
(2R)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid
Synonyms:
(2R)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid|Z-D-LEU-OH
Molecular Weight (g/mol):
265.31
Molecular Formula:
C14H19NO4
InChi Key:
USPFMEKVPDBMCG-GFCCVEGCSA-N
SMILES:
CC(C)C[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
1
–
1
of
1
results
1
N-Carbobenzoxy-D-leucine 97.0+%, TCI America™
CAS: 28862-79-5 Molecular Formula: C14H19NO4 Molecular Weight (g/mol): 265.31 MDL Number: MFCD00066068 InChI Key: USPFMEKVPDBMCG-GFCCVEGCSA-N Synonym: z-d-leu-oh,n-cbz-d-leucine,cbz-d-leu-oh,z-d-leucine,n-benzyloxycarbonyl-d-leucine,r-2-benzyloxy carbonyl amino-4-methylpentanoic acid,cbz-d-leucine,n-alpha-benzyloxycarbonyl-d-leucine,2r-4-methyl-2-phenylmethoxycarbonylamino pentanoic acid,2r-2-benzyloxy carbonyl amino-4-methylpentanoic acid PubChem CID: 7000099 IUPAC Name: (2R)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid SMILES: CC(C)C[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O