accessibility menu, dialog, popup

UserName

CAS RN 28679-16-5

H3CNCOH3CCH3NCO

CAS RN 28679-16-5

IUPAC Name: 1,6-diisocyanato-5,6-dimethylheptane
Synonyms: 1,6-diisocyanato-5,6-dimethylheptane
Molecular Weight (g/mol): 210.28
Molecular Formula: C11H18N2O2
InChi Key: VZXPHDGHQXLXJC-UHFFFAOYNA-N
SMILES: CC(CCCCN=C=O)C(C)(C)N=C=O
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)