CAS RN 28588-74-1
IUPAC Name:
2-methylfuran-3-thiol
Synonyms:
2-methyl-3-furanthiol
Molecular Weight (g/mol):
114.16
Molecular Formula:
C5H6OS
InChi Key:
RUYNUXHHUVUINQ-UHFFFAOYSA-N
SMILES:
CC1=C(S)C=CO1
1
–
1
of
1
results
1
2-Methyl-3-furanthiol 95.0+%, TCI America™
CAS: 28588-74-1 Molecular Formula: C5H6OS Molecular Weight (g/mol): 114.16 MDL Number: MFCD00010280 InChI Key: RUYNUXHHUVUINQ-UHFFFAOYSA-N Synonym: 2-methyl-3-furanthiol,3-furanthiol, 2-methyl,2-methyl-3-mercaptofuran,2-methyl-3-furanethiol,2-methyl-3-furylthiol,2-methyl-furan-3-thiol,3-mercapto-2-methylfuran,2-methyl-3-furylmercaptan,2-methyl-3-sulfanylfuran,unii-n21rw1n179 PubChem CID: 34286 IUPAC Name: 2-methylfuran-3-thiol SMILES: CC1=C(S)C=CO1