accessibility menu, dialog, popup

UserName

CAS RN 2834-79-9

IUPAC Name: 4-{[1,1'-biphenyl]-4-yl}-1,3-thiazol-2-amine
Synonyms: 4-{[1,1'-biphenyl]-4-yl}-1,3-thiazol-2-amine
Molecular Weight (g/mol): 252.34
Molecular Formula: C15H12N2S
InChi Key: HTAUVJPDFDVVHV-UHFFFAOYSA-N
SMILES: NC1=NC(=CS1)C1=CC=C(C=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results