accessibility menu, dialog, popup

UserName

ClAuClClClK

CAS RN 27988-75-6

IUPAC Name: potassium tetrachlorogoldtris(ylium)
Synonyms: potassium tetrachlorogoldtris(ylium)
Molecular Weight (g/mol): 377.86
Molecular Formula: AuCl4K
InChi Key: IKHCGJWURGSQRT-UHFFFAOYSA-J
SMILES: [K+].Cl[Au+3](Cl)(Cl)Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results