accessibility menu, dialog, popup

UserName

CAS RN 27964-99-4

H2NOOHNH2

CAS RN 27964-99-4

IUPAC Name: 2,6-diaminohexanoic acid
Synonyms: lysine lysortine α,epsilon-diaminocaproic acid lysin
Molecular Weight (g/mol): 146.19
Molecular Formula: C6H14N2O2
InChi Key: KDXKERNSBIXSRK-UHFFFAOYNA-N
SMILES: NCCCCC(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results