CAS RN 27894-50-4
IUPAC Name:
methyl (2S)-2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoate hydrochloride
Synonyms:
methyl (2S)-2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoate hydrochloride|H-LYS(Z)-OME HCL
Molecular Weight (g/mol):
330.81
Molecular Formula:
C15H23ClN2O4
InChi Key:
QPNJISLOYQGQTI-ZOWNYOTGSA-N
SMILES:
Cl.COC(=O)[C@@H](N)CCCCNC(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
N(epsilon)-Benzyloxycarbonyl-L-lysine methyl ester hydrochloride, 95%
CAS: 27894-50-4 Molecular Formula: C15H23ClN2O4 Molecular Weight (g/mol): 330.81 MDL Number: MFCD00034846 InChI Key: QPNJISLOYQGQTI-ZOWNYOTGSA-N Synonym: h-lys z-ome.hcl,h-lys z-ome hcl,s-methyl 2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride,h-lys z-ome hydrochloride,n epsilon-benzyloxycarbonyl-l-lysine methyl ester hydrochloride,nepsilon-cbz-l-lysine methyl ester hydrochloride,methyl 2s-2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride,lys z-ome.hcl,h-lys z-ome?cl,h-lys cbz-och3.hcl PubChem CID: 12935371 IUPAC Name: methyl (2S)-2-amino-6-(phenylmethoxycarbonylamino)hexanoate;hydrochloride SMILES: Cl.COC(=O)[C@@H](N)CCCCNC(=O)OCC1=CC=CC=C1