accessibility menu, dialog, popup

UserName

BFFFFBFFFFZn

CAS RN 27860-83-9

IUPAC Name: zinc(2+) bis(tetrafluoroboranuide)
Synonyms: zinc tetrafluoroborate
Molecular Weight (g/mol): 238.99
Molecular Formula: B2F8Zn
InChi Key: BUMIVIYHDVNOQE-UHFFFAOYSA-N
SMILES: [Zn++].F[B-](F)(F)F.F[B-](F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results