accessibility menu, dialog, popup

UserName

SnOOOOH3CCH3H3CH3CCH3H3C

CAS RN 2781-10-4

IUPAC Name: dibutyl[(2-ethylhexanoyl)oxy]stannyl 2-ethylhexanoate
Synonyms: dibutyl[(2-ethylhexanoyl)oxy]stannyl 2-ethylhexanoate
Molecular Weight (g/mol): 519.35
Molecular Formula: C24H48O4Sn
InChi Key: GPDWNEFHGANACG-UHFFFAOYNA-L
SMILES: CCCCC(CC)C(=O)O[Sn](CCCC)(CCCC)OC(=O)C(CC)CCCC
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)