accessibility menu, dialog, popup

UserName

ClH3CNCH3H3CSiOCH3OCH3OCH3

CAS RN 27668-52-6

IUPAC Name: dimethyl(octadecyl)[3-(trimethoxysilyl)propyl]azanium chloride
Synonyms: dimethyl(octadecyl)[3-(trimethoxysilyl)propyl]azanium chloride
Molecular Weight (g/mol): 496.29
Molecular Formula: C26H58ClNO3Si
InChi Key: WSFMFXQNYPNYGG-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)CCC[Si](OC)(OC)OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)