accessibility menu, dialog, popup

UserName

OOO(S)(R)

CAS RN 2746-19-2

IUPAC Name: (2R,6S)-4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Synonyms: (2R,6S)-4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: KNDQHSIWLOJIGP-CYDAGYADSA-N
SMILES: O=C1OC(=O)[C@@H]2C3CC(C=C3)[C@H]12
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)