accessibility menu, dialog, popup

UserName

CAS RN 274-47-5

NN

CAS RN 274-47-5

IUPAC Name: imidazo[1,5-a]pyridine
Synonyms: imidazo[1,5-a]pyridine
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: JMANUKZDKDKBJP-UHFFFAOYSA-N
SMILES: C1=NC=C2C=CC=CN12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results