accessibility menu, dialog, popup

UserName

CAS RN 2731-06-8

H3CHNH2N

CAS RN 2731-06-8

IUPAC Name: 2-(2-methyl-1H-indol-3-yl)ethan-1-amine
Synonyms: α-methyl-3-indoleethanamine
Molecular Weight (g/mol): 174.25
Molecular Formula: C11H14N2
InChi Key: CPVSLHQIPGTMLH-UHFFFAOYSA-N
SMILES: CC1=C(CCN)C2=CC=CC=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results