CAS RN 27006-76-4
IUPAC Name:
5-chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde
Synonyms:
5-chloro-1,3-dimethylpyrazole-4-carbaldehyde
Molecular Weight (g/mol):
158.59
Molecular Formula:
C6H7ClN2O
InChi Key:
SZRSMNYUEXXEBL-UHFFFAOYSA-N
SMILES:
CN1N=C(C)C(C=O)=C1Cl
1
–
1
of
1
results
1
5-Chloro-1,3-dimethylpyrazole-4-carboxaldehyde 98.0+%, TCI America™
CAS: 27006-76-4 Molecular Formula: C6H7ClN2O Molecular Weight (g/mol): 158.585 MDL Number: MFCD00052538 InChI Key: SZRSMNYUEXXEBL-UHFFFAOYSA-N Synonym: 5-chloro-1,3-dimethyl-1h-pyrazole-4-carbaldehyde,5-chloro-1,3-dimethyl-1h-pyrazole-4-carboxaldehyde,5-chloro-1,3-dimethyl pyrazole-4-aldehyde,5-chloro-1,3-dimethylpyrazole-4-carboxaldehyde,5-chloro-4-formyl-1,3-dimethylpyrazole,5-chloro-1,3-dimethyl-1h-pyrazole-4-aldehyde,1h-pyrazole-4-carboxaldehyde, 5-chloro-1,3-dimethyl,pubchem23629,acmc-20a2nm,ksc496s9t PubChem CID: 2777397 IUPAC Name: 5-chloro-1,3-dimethylpyrazole-4-carbaldehyde SMILES: CC1=NN(C(=C1C=O)Cl)C