accessibility menu, dialog, popup

UserName

NBrHNNH2

CAS RN 26944-71-8

IUPAC Name: 2-bromo-6-hydrazinylpyridine
Synonyms: 2-bromo-6-hydrazinylpyridine
Molecular Weight (g/mol): 188.03
Molecular Formula: C5H6BrN3
InChi Key: PQMFVUNERGGBPG-UHFFFAOYSA-N
SMILES: NNC1=NC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results