accessibility menu, dialog, popup

UserName

∗∗OSOOOKn

CAS RN 26837-42-3

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C2H3KO4S)n
InChi Key: —
SMILES: [K]OS(=O)(=O)OC(-*)C-*
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)