accessibility menu, dialog, popup

UserName

∗OOOO∗Mex

CAS RN 26780-50-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C3H4O2)x(C2H2O2)y
InChi Key: —
SMILES: CC(-*)C(=O)OCC(=O)O-*

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results