accessibility menu, dialog, popup

UserName

H3COBrOCH3

CAS RN 267651-23-0

IUPAC Name: 1-(3-bromo-2-methoxyphenyl)ethan-1-one
Synonyms: 1-(3-bromo-2-methoxyphenyl)ethanone
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: IBHCVANJIBGASI-UHFFFAOYSA-N
SMILES: COC1=C(C=CC=C1Br)C(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results