accessibility menu, dialog, popup

UserName

H3CCH3O

CAS RN 26465-81-6

IUPAC Name: 3,3-dimethyl-2,3-dihydro-1H-inden-1-one
Synonyms: 3,3-dimethyl-2H-inden-1-one
Molecular Weight (g/mol): 160.22
Molecular Formula: C11H12O
InChi Key: QWZAOSKLFKAEOK-UHFFFAOYSA-N
SMILES: CC1(C)CC(=O)C2=CC=CC=C12
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results