accessibility menu, dialog, popup

UserName

ClNCl

CAS RN 26452-80-2

IUPAC Name: 2,4-dichloropyridine
Synonyms: 2,4-dichloropyridine
Molecular Weight (g/mol): 147.99
Molecular Formula: C5H3Cl2N
InChi Key: TYPVHTOETJVYIV-UHFFFAOYSA-N
SMILES: ClC1=CC=NC(Cl)=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)