accessibility menu, dialog, popup

UserName

ClClOBr

CAS RN 2631-72-3

IUPAC Name: 2-bromo-1-(2,4-dichlorophenyl)ethan-1-one
Synonyms: 2-bromo-1-(2,4-dichlorophenyl)ethanone
Molecular Weight (g/mol): 267.93
Molecular Formula: C8H5BrCl2O
InChi Key: DASJDMQCPIDJIF-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C(=O)CBr)C(Cl)=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)