accessibility menu, dialog, popup

UserName

CH3ONHClCl

CAS RN 2621-62-7

IUPAC Name: N-(2,5-dichlorophenyl)acetamide
Synonyms: 2,5-dichloroacetanilide
Molecular Weight (g/mol): 204.05
Molecular Formula: C8H7Cl2NO
InChi Key: ICZFWTSENFTULW-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC(Cl)=CC=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results