accessibility menu, dialog, popup

UserName

CAS RN 261356-49-4

BNNNNK

CAS RN 261356-49-4

IUPAC Name: potassium tetracyanoboranuide
Synonyms: potassium tetracyanoboranuide
Molecular Weight (g/mol): 153.98
Molecular Formula: C4BKN4
InChi Key: SQFZXQOCCKUPEC-UHFFFAOYSA-N
SMILES: [K+].N#C[B-](C#N)(C#N)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results