accessibility menu, dialog, popup

UserName

CAS RN 2613-89-0

OHOOHO

CAS RN 2613-89-0

IUPAC Name: 2-phenylpropanedioic acid
Synonyms: 2-phenylpropanedioic acid
Molecular Weight (g/mol): 180.16
Molecular Formula: C9H8O4
InChi Key: WWYDYZMNFQIYPT-UHFFFAOYSA-N
SMILES: OC(=O)C(C(O)=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results