accessibility menu, dialog, popup

UserName

(R)(R)O(S)(R)∗OHOOHOHO∗n

CAS RN 25990-10-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C6H8O6)n
InChi Key: —
SMILES: O[C@@H]1[C@@H](O)[C@@H](O-*)[C@H](OC1-*)C(O)=O
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results