accessibility menu, dialog, popup

UserName

CAS RN 259199-65-0

IUPAC Name: N-[3-(dimethylamino)propyl]-2-[2-(4-methoxyphenyl)ethenyl]quinazolin-4-amine
Synonyms: N-[3-(dimethylamino)propyl]-2-[2-(4-methoxyphenyl)ethenyl]quinazolin-4-amine
Molecular Weight (g/mol): 362.48
Molecular Formula: C22H26N4O
InChi Key: NIHSNFSFDGHHRG-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=CC2=NC(NCCCN(C)C)=C3C=CC=CC3=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results