accessibility menu, dialog, popup

UserName

CAS RN 2589-72-2

IUPAC Name: 1-(4-hydroxyphenyl)hexan-1-one
Synonyms: 1-(4-hydroxyphenyl)hexan-1-one
Molecular Weight (g/mol): 192.26
Molecular Formula: C12H16O2
InChi Key: YDGNMJZDWLQUTE-UHFFFAOYSA-N
SMILES: CCCCCC(=O)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results