CAS RN 258521-20-9
IUPAC Name:
N'-hydroxy-2-[4-(1,3,4-oxadiazol-2-yl)phenoxy]ethanimidamide
Synonyms:
N'-hydroxy-2-[4-(1,3,4-oxadiazol-2-yl)phenoxy]ethanimidamide
Molecular Weight (g/mol):
234.22
Molecular Formula:
C10H10N4O3
InChi Key:
RNDSGEINHPCNQK-UHFFFAOYSA-N
SMILES:
NC(COC1=CC=C(C=C1)C1=NN=CO1)=NO
1
–
1
of
1
results
1
Promotions Available
2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime, 97%
2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime, 97%
CAS: 258521-20-9 Molecular Formula: C10H10N4O3 Molecular Weight (g/mol): 234.22 MDL Number: MFCD01764763 InChI Key: RNDSGEINHPCNQK-UHFFFAOYSA-N Synonym: 2-4-1,3,4-oxadiazol-2-yl phenoxy acetamidoxime,1-hydroxyimino-2-4-1,3,4-oxadiazol-2-yl phenoxy ethylamine PubChem CID: 57376339 IUPAC Name: N'-hydroxy-2-[4-(1,3,4-oxadiazol-2-yl)phenoxy]ethanimidamide SMILES: NC(COC1=CC=C(C=C1)C1=NN=CO1)=NO