accessibility menu, dialog, popup

UserName

CAS RN 25796-77-4

IUPAC Name: 3,11-dithiatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraen-7-one
Synonyms: 3,11-dithiatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraen-7-one
Molecular Weight (g/mol): 192.25
Molecular Formula: C9H4OS2
InChi Key: HFIUHKXJUKKOIZ-UHFFFAOYSA-N
SMILES: O=C1C2=C(SC=C2)C2=C1C=CS2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results