CAS RN 25639-42-3
CAS RN 25639-42-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
115.2
Molecular Formula:
C7H15O
InChi Key:
—
SMILES:
C*.OC1CCCCC1
1
–
1
of
1
results
1
Methylcyclohexanol (2-,3-,4- and cis-,trans- mixture) 88.0+%, TCI America™
CAS: 25639-42-3 Molecular Formula: C7H15O Molecular Weight (g/mol): 115.20 MDL Number: MFCD01778225