CAS RN 2557-81-5
CAS RN 2557-81-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Phenylsulfur Pentafluoride 98.0+%, TCI America™
CAS: 2557-81-5 Molecular Formula: C6H5F5S Molecular Weight (g/mol): 204.158 MDL Number: MFCD09263740 InChI Key: DMNYFEWFSFTYEL-UHFFFAOYSA-N PubChem CID: 9942464 IUPAC Name: pentafluoro(phenyl)-$l^{6}-sulfane SMILES: C1=CC=C(C=C1)S(F)(F)(F)(F)F