accessibility menu, dialog, popup

UserName

OHBHOHOO

CAS RN 25487-66-5

IUPAC Name: 3-(dihydroxyboranyl)benzoic acid
Synonyms: 3-(dihydroxyboranyl)benzoic acid
Molecular Weight (g/mol): 165.94
Molecular Formula: C7H7BO4
InChi Key: DBVFWZMQJQMJCB-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC(=C1)C(O)=O
Physical Form 
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)