accessibility menu, dialog, popup

UserName

H3COO

CAS RN 25414-22-6

IUPAC Name: 2-methoxyfuran
Synonyms: 2-methoxyfuran
Molecular Weight (g/mol): 98.1
Molecular Formula: C5H6O2
InChi Key: OXCGHDNCMSOEBZ-UHFFFAOYSA-N
SMILES: COC1=CC=CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)