accessibility menu, dialog, popup

UserName

H3COOOHOCH3O

CAS RN 25395-31-7

IUPAC Name: 1-(acetyloxy)-3-hydroxypropan-2-yl acetate
Synonyms: 1,2,3-propanetriol, diacetate diacetin acetin, 1,2-di-
Molecular Weight (g/mol): 176.17
Molecular Formula: C7H12O5
InChi Key: UXDDRFCJKNROTO-UHFFFAOYNA-N
SMILES: CC(=O)OCC(CO)OC(C)=O
Boiling Point 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)