accessibility menu, dialog, popup

UserName

NNN

CAS RN 25373-49-3

IUPAC Name: 2-(1H-imidazol-1-yl)benzonitrile
Synonyms: 2-(imidazol-1-yl)benzonitrile
Molecular Weight (g/mol): 169.19
Molecular Formula: C10H7N3
InChi Key: MNKBJOSIVSQUBI-UHFFFAOYSA-N
SMILES: N#CC1=CC=CC=C1N1C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)