accessibility menu, dialog, popup

UserName

H3CCH3H3CSH

CAS RN 25360-09-2

IUPAC Name: 13,13-dimethyltetradecane-1-thiol
Synonyms: 13,13-dimethyltetradecane-1-thiol
Molecular Weight (g/mol): 258.51
Molecular Formula: C16H34S
InChi Key: KZIYAWAYDPPPIU-UHFFFAOYSA-N
SMILES: CC(C)(C)CCCCCCCCCCCCS

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results