accessibility menu, dialog, popup

UserName

∗∗NOn

CAS RN 25249-54-1

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C6H9NO)n
InChi Key: —
SMILES: *-CC(-*)N1CCCC1=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)