accessibility menu, dialog, popup

UserName

CAS RN 25121-81-7

IUPAC Name: 2-bromothieno[2,3-b]thiophene
Synonyms: 2-bromothieno[2,3-b]thiophene
Molecular Weight (g/mol): 219.11
Molecular Formula: C6H3BrS2
InChi Key: DRHZNSHNAFMJJF-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(SC=C2)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results