accessibility menu, dialog, popup

UserName

CHN

CAS RN 2510-22-7

IUPAC Name: 4-ethynylpyridine
Synonyms: 4-ethynylpyridine
Molecular Weight (g/mol): 103.12
Molecular Formula: C7H5N
InChi Key: FDEDJRHULYIJOR-UHFFFAOYSA-N
SMILES: C#CC1=CC=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)